Anti-Uroguanylin Rabbit Polyclonal Antibody

Supplier: Proteintech
18113-1-AP-150UL
10096-818EA 402 USD
10096-818
Anti-Uroguanylin Rabbit Polyclonal Antibody
Antibodies
Uroguanylin, also named as GUCA2B, is a candidate intestinal natriuretic hormone that controls the sodium and water balance between the intestine and the kidneys. It has been reported that the uroguanylin knockout mouse exhibits elevated blood pressure. Therefore, the GUCA2B gene is thought to be a susceptibility gene for essential hypertension (EH).

Western Blot: Mouse pancreas Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: Uroguanylin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR