Anti-IL20 Rabbit Polyclonal Antibody

Supplier: Proteintech
18104-1-AP-150UL
10088-806EA 402 USD
10088-806
Anti-IL20 Rabbit Polyclonal Antibody
Antibodies
Interleukin-20 (IL-20) is a protein belonging to the IL-10 family of cytokines. IL-20 receptor (IL-20R) and IL-20 are expressed in several normal tissue types, including the lung, skin, and kidney. IL-20 is a cytokine structurally related to interleukin 10. IL-20 is produced by activated keratinocytes and monocytes and transmits an intracellular signal through two distinct cell-surface receptor complexes on keratinocytes and other epithelial cells. IL-20 can stimulate STAT3 activation in keratinocytes. IL-20 regulates proliferation and differentiation of keratinocytes during inflammation, particularly inflammation associated with the skin. In addition, IL-20 also causes cell expansion of multipotential hematopoietic progenitor cells. IL-20 is highly associated with potent inflammatory diseases such as psoriasis, contact hypersensitivity, rheumatoid arthritis, and atherosclerosis.

Western Blot:Transfected HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: IL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR