Anti-CLOCK Rabbit Polyclonal Antibody

Supplier: Proteintech
18094-1-AP-150UL
10084-874EA 402 USD
10084-874
Anti-CLOCK Rabbit Polyclonal Antibody
Antibodies
Circadian locomoter output cycles protein kaput (CLOCK), also named as BHLHE8 or KIAA0334, is a 846 amino acid protein, which contains one bHLH domain, one PAC domain, and two PAS domains. CLOCK localizes in the nucleus and cytoplasm. CLOCK) is expressed in all tissues examined including spleen, thymus, prostate, testis, ovary, small intestine, colon, leukocytes, heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. ARNTL/2-CLOCK heterodimers activate E-box element (5'-CACGTG-3') transcription of a number of proteins of the circadian clock, such as PER1 and PER2. The calculated molecular weight of CLOCK is 95kDa, but we detected a 150 kDa protein by western blot. Our result is similar to the result of reseach paper .

Western Blot: HeLa Cells, 1:500-1:5000; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: CLOCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR