Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18084-1-AP-150UL
10091-344EA 402 USD
10091-344
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:1000-1:10000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: NTF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR