Anti-USP49 Rabbit Polyclonal Antibody

Supplier: Proteintech
18066-1-AP-150UL
10096-886EA 402 USD
10096-886
Anti-USP49 Rabbit Polyclonal Antibody
Antibodies
USPs are cysteine proteases that vary greatly in size and structural complexity. Outside of the core catalytic domain, USPs exhibit N-terminal and C-terminal extensions that have been proposed to play roles in determining cellular localization and substrate specificity of these enzymes. USP49(Ubiquitin carboxyl-terminal hydrolase 49) belongs to this family. It has 2 isoforms produced by alternative splicing.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: USP49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR