Anti-PTPN14 Rabbit Polyclonal Antibody

Supplier: Proteintech
18053-1-AP-150UL
10093-056EA 402 USD
10093-056
Anti-PTPN14 Rabbit Polyclonal Antibody
Antibodies
PTPN14 (tyrosine-protein phosphatase non-receptor type 14) is also named as PEZ and belongs to the protein-tyrosine phosphatase family and non-receptor class subfamily. It is a 130-kDa multi-domain protein expressed in breast, kidney, skeletal muscle, lung, placenta, and characterized by an N-terminal FERM domain and a C-terminal PTP domain with an intervening sequence containing an acidic region and a putative SH3 domain-binding sequence . Defects in PTPN14 are a cause of choanal atresia and lymphedema (CHATLY).

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: PTPN14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR