Anti-fetuin-B Rabbit Polyclonal Antibody

Supplier: Proteintech
18052-1-AP-150UL
10086-954EA 402 USD
10086-954
Anti-fetuin-B Rabbit Polyclonal Antibody
Antibodies
Fetuin-B is a protein that in humans is encoded by the FETUB gene. The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. It may be a protease inhibitor required for egg fertilization.

Western Blot: Human Blood Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: fetuin-B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR