Anti-DNA Ligase I Rabbit Polyclonal Antibody

Supplier: Proteintech
18051-1-AP-150UL
10085-580EA 524.48 USD
10085-580
Anti-DNA Ligase I Rabbit Polyclonal Antibody
Antibodies
LIG1, also named as LRIG1, acts as a feedback negative regulator of signaling by receptor tyrosine kinases, through a mechanism that involves enhancement of receptor ubiquitination and accelerated intracellular degradation.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: DNA Ligase I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR