Anti-MLN51/CASC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18047-1-AP-150UL
10090-280EA 524.48 USD
10090-280
Anti-MLN51/CASC3 Rabbit Polyclonal Antibody
Antibodies
CASC3(cancer susceptibility candidate 3) is also named as MLN51 and belongs to the CASC3 family. It is induced by CSF2, and is important in the proliferation of RA fibroblast-like synoviocytes in the pathogenesis of rheumatoid arthritis (RA). It is a nucleocytoplasmic protein containing, within its aminoterminal half, a coiled-coil domain followed by two nuclear localization signals (NLSs) responsible for nuclear localization and, in its carboxyl-terminal half, a nuclear export signal(NES) that mediates cytoplasmic retention. It possesses many potential phosphorylation sites and is found to be phosphorylated in both normal and cancer-derived cells.. This antibody is specific to CASC3.

Western Blot:A2780 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: MLN51/CASC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR