Anti-EDAR Rabbit Polyclonal Antibody

Supplier: Proteintech
18032-1-AP-150UL
10086-252EA 402 USD
10086-252
Anti-EDAR Rabbit Polyclonal Antibody
Antibodies
EDAR, also named as DL, is a receptor for EDA isoform A1, but not for EDA isoform A2. It mediates the activation of NF-kappa-B and JNK. EDAR may promote caspase-independent cell death. It is expressed in adjacent or partially overlapping layers in the developing human skin with MW 48 kDa.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: EDAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR