Anti-CGNL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18031-1-AP-150UL
10084-416EA 402 USD
10084-416
Anti-CGNL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:1000-1:10000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CGNL1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR