Anti-TRPM5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18027-1-AP-150UL
10095-924EA 402 USD
10095-924
Anti-TRPM5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Colon Tissue, 1:500-1:5000; IF: Mouse olfactory epithelium Tissue, N/A

Type: Primary
Antigen: TRPM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR