Anti-ICA1L Rabbit Polyclonal Antibody

Supplier: Proteintech
18021-1-AP-150UL
10081-216EA 402 USD
10081-216
Anti-ICA1L Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Brain, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: ICA1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR