Anti-EIF2B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18010-1-AP-150UL
10086-350EA 537.7 USD
10086-350
Anti-EIF2B1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:500-1:5000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: EIF2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR