Anti-MME Rabbit Polyclonal Antibody

Supplier: Proteintech
18008-1-AP-150UL
10081-630EA 402 USD
10081-630
Anti-MME Rabbit Polyclonal Antibody
Antibodies
MME is also names as neprilysin,CALLA,NEP,SFE,CD10 and belongs to the peptidase M13 family.It is a glycoprotein that is particularly abundant in kidney, where it is present on the brush border of proximal tubules and on glomerular epithelium.CD10 has been considered a useful marker in the diagnosis of renal carcinomas, because of its expression in clear cell and papillary renal cell carcinomas and its absence in chromophobe renal cell carcinomas.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IP: Raji Cells, 1:200-1:2000; FC:Jurkat Cells, N/A

Type: Primary
Antigen: MME
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR