Anti-APOA4 Rabbit Polyclonal Antibody

Supplier: Proteintech
17996-1-AP-150UL
10083-008EA 402 USD
10083-008
Anti-APOA4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Blood Tissue, 1:500-1:5000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: APOA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR