Anti-ACHE Rabbit Polyclonal Antibody

Supplier: Proteintech
17975-1-AP-150UL
10082-422EA 402 USD
10082-422
Anti-ACHE Rabbit Polyclonal Antibody
Antibodies
The classical role of acetylcholinesterase (AChE) is to terminate cholinergic neurotransmission by rapid hydrolysis of the neurotransmitter acetylcholine (ACh) at cholinergic synapses and neuromuscular junctions. AChE activity is significantly lower in aspirates from squamous cell carcinomas and the molecular distribution of AChE in both bronchial cells and fluids consisted of amphiphilic monomers and dimers. It has 4 isoforms produced by alternative splicing.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR