Anti-RGS9 Rabbit Polyclonal Antibody

Supplier: Proteintech
17970-1-AP-150UL
10093-634EA 524.48 USD
10093-634
Anti-RGS9 Rabbit Polyclonal Antibody
Antibodies
RGS9 is a member of the RGS family of signaling proteins that suppress the activity of G proteins by promoting their deactivation. There are two splice isoforms of RGS9 with quite different properties and patterns of expression. RGS9-1 is mainly found in the eye and is involved in regulation of phototransduction in rod and cone cells of the retina, while RGS9-2 is is expressed specifically in the striatum, a brain region involved in controlling movement, motivation, mood and addition. This antibody got about 58kD in the mouse eye and 70kD in the mouse brain in western blotting, both in line with their expected masses. Defects in RGS9 are a cause of prolonged electroretinal response suppression (PERRS).

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: RGS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR