Anti-LOX Rabbit Polyclonal Antibody

Supplier: Proteintech
17958-1-AP-150UL
10089-760EA 402 USD
10089-760
Anti-LOX Rabbit Polyclonal Antibody
Antibodies
LOX is a member of a multigene family (LOX, LOXL, LOXL2, LOXL3, and LOXL4). Lysyl oxidase (LOX), a copper-dependent enzyme, plays a critical role in the formation and repair of the extracellularmatrix (ECM) by catalyzing the crosslinking of elastin and collagen. LOX expression appears to be upregulated in malignancies and has recently been suggested to play a role in regulating tumor metastasis. It is synthesized as a 50 kDa proenzyme, secreted, and processed into an 32 kDa mature, active enzyme and an 18-kDa propeptide.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200; IP: Mouse Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR