Anti-PD-L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17952-1-AP-150UL
10091-838EA 537.7 USD
10091-838
Anti-PD-L1 Rabbit Polyclonal Antibody
Antibodies
Programmed cell death ligand 1(CD274, or B7-H1, PD-L1), is the first member of B7 family to be discovered. B7 family molecules are type I transmembrane proteins belonging to the immunoglobulin superfamily. In concert with their CD28 family receptors, the B7s are key regulators of the adaptive immune response. CD274 is suggested a negative regulator of T and B cell, and play important role in mediating tolerance of lymphocytes to self-antigens. It also involved in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.

Western Blot: Mouse Heart Tissue, 1:200-1:1000; IHC: Mouse Heart Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:10-1:100

Type: Primary
Antigen: PD-L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR