Anti-GLRA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17951-1-AP-150UL
10087-586EA 402 USD
10087-586
Anti-GLRA1 Rabbit Polyclonal Antibody
Antibodies
Glycine is an inhibitory neurotransmitter in the central nervous system. The glycine receptor (GlyR) is a member of the nicotinic acetylcholine receptor of ligand-gated ion channels. This receptor has important roles in a variety of physiological processes, especially in mediating inhibitory neurotransmission in the spinal cord and brain stem. GlyR is a pentameric receptor comprised of alpha and beta subunits. Four alpha-subunits (GLRA1, GLRA2, GLRA3, GLRA4) and one beta-subunit (GLRB) of GlyR have been identified.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: GLRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR