Anti-PRR5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17948-1-AP-150UL
10092-848EA 402 USD
10092-848
Anti-PRR5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: PRR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR