Anti-PTPRR Rabbit Polyclonal Antibody

Supplier: Proteintech
17937-1-AP-150UL
10092-332EA 402 USD
10092-332
Anti-PTPRR Rabbit Polyclonal Antibody
Antibodies
PTPRR(protein tyrosine phosphatase, receptor type, R) is also named as PCPTP1, PTP-SL, PTPBR7 and belongs to the protein-tyrosine phosphatase family. It sequesters mitogen-activated protein kinases (MAPKs) such as MAPK1, MAPK3 and MAPK14 in the cytoplasm in an inactive form. It has four isoforms that they have molecular weights of 72, 60, 42 and 37 kDa, respectively, and differ in the length of their N-terminal part which result in distinct subcellular localizations. All PTPRR isoforms are found to be short-lived proteins, with half-lives of around 3-5 hours, and are constitutively phosphorylated on one or two of the previously identified protein kinase A and MAP kinase target sites.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: PTPRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR