Anti-CRYGC Rabbit Polyclonal Antibody

Supplier: Proteintech
17931-1-AP-150UL
10085-224EA 402 USD
10085-224
Anti-CRYGC Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: CRYGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR