Anti-ETFB Rabbit Polyclonal Antibody

Supplier: Proteintech
17925-1-AP-150UL
10086-676EA 402 USD
10086-676
Anti-ETFB Rabbit Polyclonal Antibody
Antibodies
Electron transfer flavoprotein (ETF) exists in the mitochondrial matrix as a heterodimer of 30-kD alpha subunits (ETFA)and 28-kD beta subunits (ETFB) and contains 1 flavin adenine dinucleotide (FAD) and 1 adenosine 5-prime monophosphate (AMP) per heterodimer.It serves as a specific electron acceptor for several dehydrogenases, including five acyl-CoA dehydrogenases, glutaryl-CoA and sarcosine dehydrogenase.ETFB has 2 isoforms produced by alternative splicing.Defects in ETFB are the cause of glutaric aciduria type 2B (GA2B).

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ETFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR