Anti-MDM4 Rabbit Polyclonal Antibody

Supplier: Proteintech
17914-1-AP-150UL
10090-072EA 537.7 USD
10090-072
Anti-MDM4 Rabbit Polyclonal Antibody
Antibodies
MDM4 is also named as MDMX. It inhibits the activity of the tumor suppressor p53 by primarily cooperating with the p53 feedback regulator MDM2. MDM4 resides mostly in the cytoplasm, but can be recruited to the nucleus by MDM2.. The predicted mass of the protein is 54 kD, while the observed mass is 80 kD, a difference which stated is probably due to phosphorylation or other posttranslational modification. It can be ubiquitinated and degraded by MDM2. It has 5 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: MDM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR