Anti-EPO Rabbit Polyclonal Antibody

Supplier: Proteintech
17908-1-AP-150UL
10086-588EA 402 USD
10086-588
Anti-EPO Rabbit Polyclonal Antibody
Antibodies
Erythropoietin (Epo) is a member of the EPO/TPO family and encodes a secreted, glycosylated cytokine composed of four alpha helical bundles. The protein is found in the plasma and regulates red cell production by promoting erythroid differentiation and initiating hemoglobin synthesis. Its effect is realized by binding erythropoietin receptor (EpoR) expressed on erythroid progenitor cells. EpoR, is a glycoprotein expressed on megakaryocytes, erythroid progenitors and endothelial cells. Epo also has neuroprotective activity against a variety of potential brain injuries and antiapoptotic functions in several tissue types. In western blotting, we got three bands 37 kDa, 45 kDa and 80 kDa. We are unsure as to the identity the extra bands of 45 kDa and 80 kDa.

IHC: Human Kidney Tissue, 1:20-1:200; Western Blot: Mouse Liver Tissue, 1:200-1:1000

Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR