Anti-BHLHE40 Rabbit Polyclonal Antibody

Supplier: Proteintech
17895-1-AP-150UL
10083-594EA 402 USD
10083-594
Anti-BHLHE40 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:200-1:2000

Type: Primary
Antigen: BHLHE40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR