Anti-MPV17L Rabbit Polyclonal Antibody

Supplier: Proteintech
17884-1-AP-150UL
10090-396EA 402 USD
10090-396
Anti-MPV17L Rabbit Polyclonal Antibody
Antibodies
MpV17 mitochondrial membrane protein-like protein (MPV17L) is a protein with high sequence homology with MPV17. MPV17L was initially identified as a peroxisomal membrane protein which is involved in the development of early-onset of glemerulosclerosis. But now its localization is still controversial. Human MPV17L has two variants ubiquitously expressed in human tissues, but with different cellular localizations, and plays roles in regulating expression of antioxidant enzymes. Catlog#17884-1-AP is a rabbit polyclonal antibody raised against the full-length of human MPV17L.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: MPV17L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR