Anti-LIN9 Rabbit Polyclonal Antibody

Supplier: Proteintech
17882-1-AP-150UL
10089-704EA 537.7 USD
10089-704
Anti-LIN9 Rabbit Polyclonal Antibody
Antibodies
LIN9, also named as BARA or TGS, is a 542 amino acid protein, which belongs to the lin9 family. LIN9 localizes in the nucleus and is expressed in thymus and testis. LIN9 acts as a tumor suppressor and inhibits DNA synthesis. LIN9 plays a role in the expression of genes, which required for the G1/S transition.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: LIN9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR