Anti-CAMSAP1L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17880-1-AP-150UL
10084-044EA 402 USD
10084-044
Anti-CAMSAP1L1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: CAMSAP1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR