Anti-CD226 Rabbit Polyclonal Antibody

Supplier: Proteintech
17842-1-AP-150UL
10084-300EA 402 USD
10084-300
Anti-CD226 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD226
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR