Anti-MGAT4C Rabbit Polyclonal Antibody

Supplier: Proteintech
17841-1-AP-150UL
10090-182EA 402 USD
10090-182
Anti-MGAT4C Rabbit Polyclonal Antibody
Antibodies
MGAT4C is predicted to be a glycosyltransferase that catalyzes a GlcNAc β1-4 linkage to the core mannose residues of N-glycans, resulting in N-glycans with tri- or tetraantennary structures.Recently MGAT4C has been found to get involved in processing complex N-glycan structures on CD133 and CD133 has been identified as a substrate of MGAT4C. Given to the fact that CD133 is widely used as a marker of numerous stem cell and cancer stem cell types, MGAT4C may have an important role in certain cancers.(21937449)

Western Blot:SH-SY5Y Cells, 1:500-1:5000

Type: Primary
Antigen: MGAT4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR