Anti-IL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17835-1-AP-150UL
10088-834EA 402 USD
10088-834
Anti-IL3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Recombinant Protein, 1:1000-1:10000

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR