Anti-SPERT Rabbit Polyclonal Antibody

Supplier: Proteintech
17826-1-AP-150UL
10095-088EA 402 USD
10095-088
Anti-SPERT Rabbit Polyclonal Antibody
Antibodies
SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins . Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: SPERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR