Anti-CACNA1G Rabbit Polyclonal Antibody

Supplier: Proteintech
17821-1-AP-150UL
10083-936EA 402 USD
10083-936
Anti-CACNA1G Rabbit Polyclonal Antibody
Antibodies
IF: Mouse Heart Tissue, 1:20-1:200

Type: Primary
Antigen: CACNA1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR