Anti-RABL2A Rabbit Polyclonal Antibody

Supplier: Proteintech
17816-1-AP-150UL
10093-254EA 402 USD
10093-254
Anti-RABL2A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse pancreas Tissue, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: RABL2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR