Anti-COQ10A Rabbit Polyclonal Antibody

Supplier: Proteintech
17812-1-AP-150UL
10085-020EA 402 USD
10085-020
Anti-COQ10A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: COQ10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR