Anti-PGK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17811-1-AP-150UL
10092-148EA 402 USD
10092-148
Anti-PGK1 Rabbit Polyclonal Antibody
Antibodies
The PGK1 gene encodes phosphoglycerate kinase-1, also known as ATP:3-phosphoglycerate 1-phosphotransferase (EC 2.7.2.3), which catalyzes the reversible conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate during glycolysis, generating one molecule of ATP. It Belongs to the phosphoglycerate kinase family and defects in PGK1 are the cause of phosphoglycerate kinase 1 deficiency (PGK1D).

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: PGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR