Anti-c-Myb Rabbit Polyclonal Antibody

Supplier: Proteintech
17800-1-AP-150UL
10084-906EA 402 USD
10084-906
Anti-c-Myb Rabbit Polyclonal Antibody
Antibodies
The MYB protein is a transcription factor that plays a key role in regulating the proliferation/differentiation of progenitor cells in bone marrow, colonic crypts, and neurogenic niches . It is involved in the regulation of epithelial-to-mesenchymal transition (EMT) and invasion in neuroblastoma, colon carcinoma, and embryonic kidney cells through the upregulation of the transcription repressor Slug . It also a DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3', and plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.

Western Blot: HL-60 Cells, 1:200-1:1000

Type: Primary
Antigen: c-Myb
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR