Anti-SLC25A33 Rabbit Polyclonal Antibody

Supplier: Proteintech
17794-1-AP-150UL
10094-758EA 402 USD
10094-758
Anti-SLC25A33 Rabbit Polyclonal Antibody
Antibodies
Western Blot:U2Os Cells, 1:200-1:2000

Type: Primary
Antigen: SLC25A33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR