Anti-PDE7B Rabbit Polyclonal Antibody

Supplier: Proteintech
17771-1-AP-150UL
10092-014EA 402 USD
10092-014
Anti-PDE7B Rabbit Polyclonal Antibody
Antibodies
PDE7B(cAMP-specific 3',5'-cyclic phosphodiesterase 7B) belongs to the cyclic nucleotide phosphodiesterase family and PDE7 subfamily,which regulate intracellular levels of cAMP and cGMP, cyclic nucleotides that play important roles as second messengers in multiple physiologic processes. PDE7B gene is expressed predominantly in the brain and in a number of other tissues including liver, heart, thyroid glands, and skeletal muscle, but not expressed in blood leukocytes .

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: PDE7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR