Anti-CORO1A Rabbit Polyclonal Antibody

Supplier: Proteintech
17760-1-AP-150UL
10085-030EA 402 USD
10085-030
Anti-CORO1A Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: CORO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR