Anti-DCK Rabbit Polyclonal Antibody

Supplier: Proteintech
17758-1-AP-150UL
10085-696EA 402 USD
10085-696
Anti-DCK Rabbit Polyclonal Antibody
Antibodies
Deoxycytidine kinase (dCK) is required for the phosphorylation of several deoxyribonucleoside analogues that are widely employed as chemotherapeutic agents. It is also a key enzyme in the phosphorylation of a variety of antineoplastic and antiviral nucleoside analogs including cytosine arabinoside (araC) and dideoxycytidine (ddCyd); deficiency of deoxycytidine kinase activity mediates resistance to these drugs. This protein can exsit as a homodimer.

Western Blot: MCF7 Cells, 1:500-1:5000

Type: Primary
Antigen: DCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR