Anti-TJAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17734-1-AP-150UL
10096-034EA 402 USD
10096-034
Anti-TJAP1 Rabbit Polyclonal Antibody
Antibodies
TJAP1, also named as PILT and TJP4, is colocalized at the tight junctions. TJAP1 has some forms with MW 60 kDa (native) and 86-90 kDa (modified).

Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000

Type: Primary
Antigen: TJAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR