Anti-ATP6V1F Rabbit Polyclonal Antibody

Supplier: Proteintech
17725-1-AP-150UL
10083-376EA 402 USD
10083-376
Anti-ATP6V1F Rabbit Polyclonal Antibody
Antibodies
ATP6V1F(V-type proton ATPase subunit F) is also named as ATP6S14, VATF and belongs to the V-ATPase F subunit family. It generates an electrochemical proton gradient that is acid and positive inside synaptic vesicles. ATP6V1F plays a major role as energizers of animal plasma membranes, especially apical plasma membranes of epithelial cells. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 14 kDa and 16 kDa.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: ATP6V1F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR