Anti-PRX5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17724-1-AP-150UL
10092-760EA 402 USD
10092-760
Anti-PRX5 Rabbit Polyclonal Antibody
Antibodies
Prx5(Peroxiredoxin-5, mitochondrial) reduces hydrogen peroxide and alkyl hydroperoxides with reducing equivalents provided through the thioredoxin system. While PRDX5 was localized to fibroblasts in normal tendon, it was localized to fibroblasts and endothelial cells in degenerative tendon.It can form a dimer of 36 kDa.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: PRX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR