Anti-MPP5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17710-1-AP-150UL
10090-380EA 402 USD
10090-380
Anti-MPP5 Rabbit Polyclonal Antibody
Antibodies
MPP5, also named as PALS1, belongs to the MAGUK family which interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. MPP5 is thought to organize intracellular PALS1-CRB-MUPP1 protein scaffolds in the retina that are involved in maintenance of photoreceptor–Mu¨ller glia cell adhesion. MPP5 plays a role in tight junctions biogenesis and in the establishment of cell polarity in epithelial cells.

Western Blot:Y79 Cells, 1:500-1:5000; IP: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MPP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR