Anti-USP28 Rabbit Polyclonal Antibody

Supplier: Proteintech
17707-1-AP-150UL
10096-860EA 524.48 USD
10096-860
Anti-USP28 Rabbit Polyclonal Antibody
Antibodies
USP28(Ubiquitin carboxyl-terminal hydrolase 28) is also named as KIAA1515. USP28 is a 53BP1-interacting protein and a putative deubiquitinating enzyme, and belongs to a family of enzymes, called deubiquitinating enzymes (DUBs), which remove ubiquitin from the ubiquitin conjugates. It can be phosphorylated upon DNA damage at Ser-67 and Ser-714, by ATM or ATR. This protein has 2 isoforms produced by alternative splicing with the molecular mass of 122 kDa and 119 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: USP28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR