Anti-CDCA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17701-1-AP-150UL
10084-500EA 402 USD
10084-500
Anti-CDCA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: CDCA2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR